Assay Details
Target Gene Details
Entrez Gene ID: | 419 |
Gene Name: | ADP-ribosyltransferase 3 |
Gene Aliases: |
ARTC3 |
Location: |
Chr.4:76011180-76112802 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ART3 | NM_001130017.2 | NP_001123489.1 | ||
| AK225860.1 | ||||
| AY543656.1 | AAT10278.1 | |||
| AY543657.1 | AAT10279.1 | |||
| AY543658.1 | AAT10280.1 | |||
| AY543659.1 | AAT10281.1 | |||
| DB451386.1 | ||||
| U47054.1 | AAB01894.1 |
Target Gene Details
Entrez Gene ID: | 6373 |
Gene Name: | C-X-C motif chemokine ligand 11 |
Gene Aliases: |
H174, I-TAC, IP-9, IP9, SCYB11, SCYB9B, b-R1 |
Location: |
Chr.4:76033682-76036197 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL11 | NM_001302123.1 | 4 | 617 | NP_001289052.1 |
| NM_005409.4 | 4 | 621 | NP_005400.1 | |
| AF002985.1 | 4 | 498 | AAC51845.1 | |
| AF030514.1 | 4 | 480 | AAC39775.1 | |
| AL703031.1 | ||||
| BC005292.1 | 4 | 471 | AAH05292.1 | |
| BC012532.1 | 4 | 625 | AAH12532.1 | |
| BC110986.1 | 4 | 480 | AAI10987.1 | |
| BP289021.1 | 4 | 523 | ||
| U59286.1 | AAB17374.1 | |||
| U66096.1 | 4 | 456 | AAD38867.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27112 | Chr.4:76027416 - 76056627 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map