Assay Details
Target Gene Details
Entrez Gene ID: | 10153 |
Gene Name: | CCAAT/enhancer binding protein zeta |
Gene Aliases: |
CBF, CBF2, HSP-CBF, NOC1 |
Location: |
Chr.2:37201629-37231597 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPZ | NM_005760.2 | NP_005751.2 | ||
| AK312322.1 | ||||
| BC034475.2 | AAH34475.1 | |||
| M37197.1 | AAA51924.1 |
Target Gene Details
Entrez Gene ID: | 100505876 |
Gene Name: | CEBPZ opposite strand |
Gene Aliases: |
CEBPZ-AS1 |
Location: |
Chr.2:37196451-37216193 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPZOS | NR_136316.1 | |||
| BX093544.1 | ||||
| HY189314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2684 | Chr.2:37203356 - 37236290 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map