Assay Details
Target Gene Details
Entrez Gene ID: | 401541 |
Gene Name: | centromere protein P |
Gene Aliases: |
CENP-P |
Location: |
Chr.9:92325457-92615164 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPP | NM_001012267.2 | NP_001012267.1 | ||
| NM_001286969.1 | NP_001273898.1 | |||
| XM_011518689.1 | XP_011516991.1 | |||
| AK092603.1 | ||||
| AK307435.1 | ||||
| BC071726.1 | AAH71726.1 | |||
| BX537851.1 | CAD97859.1 |
Target Gene Details
Entrez Gene ID: | 4958 |
Gene Name: | osteomodulin |
Gene Aliases: |
OSAD, SLRR2C |
Location: |
Chr.9:92414245-92424554 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OMD | NM_005014.2 | 2 | 739 | NP_005005.1 |
| AB000114.1 | 2 | 470 | BAA19055.1 | |
| AK225922.1 | 2 | 584 | ||
| AK313055.1 | 2 | 577 | ||
| AY358872.1 | 2 | 637 | AAQ89231.1 | |
| BC046356.1 | 2 | 640 | AAH46356.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053205 | Chr.9:92356913 - 92420330 on Build GRCh38 | Loss |
|
| nsv1049599 | Chr.9:92407857 - 92503276 on Build GRCh38 | Loss |
|
| esv3891693 | Chr.9:92163398 - 92538277 on Build GRCh38 | Gain |
|
| nsv1047337 | Chr.9:92343356 - 92585202 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map