Assay Details
Target Gene Details
Entrez Gene ID: | 6307 |
Gene Name: | methylsterol monooxygenase 1 |
Gene Aliases: |
DESP4, ERG25, MCCPD, SC4MOL |
Location: |
Chr.4:165327666-165343164 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSMO1 | NM_001017369.2 | 5 | 1613 | NP_001017369.1 |
| NM_006745.4 | 6 | 1899 | NP_006736.1 | |
| XM_005263176.2 | 6 | 2016 | XP_005263233.1 | |
| BC010653.1 | 5 | 1793 | AAH10653.1 | |
| BC107879.1 | 6 | 1861 | AAI07880.1 | |
| BU676028.1 | 1 | 343 |
More Information
Additional Information:
For this assay, SNP(s) [rs140707692] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map