Assay Details
Target Gene Details
Entrez Gene ID: | 5433 |
Gene Name: | RNA polymerase II subunit D |
Gene Aliases: |
HSRBP4, HSRPB4, RBP4, RPB16 |
Location: |
Chr.2:127846266-127858155 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2D | NM_004805.3 | NP_004796.1 | ||
| BC017205.2 | AAH17205.1 | |||
| BC093795.1 | AAH93795.1 | |||
| BC093797.1 | AAH93797.1 | |||
| U85510.1 | AAC80226.1 |
More Information
Set Membership: |
|

Genomic Map