Assay Details
Target Gene Details
Entrez Gene ID: | 55117 |
Gene Name: | solute carrier family 6 member 15 |
Gene Aliases: |
NTT73, SBAT1, V7-3, hv7-3 |
Location: |
Chr.12:84859488-84912876 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A15 | NM_001146335.2 | 11 | 2640 | NP_001139807.1 |
| NM_182767.5 | 12 | 3117 | NP_877499.1 | |
| AF351619.1 | 6 | 1657 | AAN76519.1 | |
| AK001178.1 | 7 | 1801 | BAA91536.1 | |
| AL050066.1 | 1 | 621 | ||
| BC070040.1 | 12 | 3081 | AAH70040.1 | |
| CR749673.1 | 11 | 2595 | CAH18464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035236 | Chr.12:84396728 - 84956654 on Build GRCh38 | Loss |
|
| dgv2761n54 | Chr.12:84830362 - 84945475 on Build GRCh38 | Loss |
|
| nsv559644 | Chr.12:84791617 - 84863666 on Build GRCh38 | Loss |
|
| dgv208n27 | Chr.12:84855439 - 84945475 on Build GRCh38 | Loss |
|
| nsv1040452 | Chr.12:84785807 - 85354129 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map