Assay Details
Target Gene Details
Entrez Gene ID: | 149069 |
Gene Name: | doublecortin domain containing 2B |
Gene Aliases: |
- |
Location: |
Chr.1:32208813-32216196 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCDC2B | NM_001099434.1 | NP_001092904.1 | ||
| BM980133.1 |
Target Gene Details
Entrez Gene ID: | 56063 |
Gene Name: | transmembrane protein 234 |
Gene Aliases: |
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7 |
Location: |
Chr.1:32204807-32222371 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM234 | NM_019118.4 | NP_061991.3 | ||
| NR_133634.1 | ||||
| NR_133635.1 | ||||
| NR_133636.1 | ||||
| XM_011541795.2 | 7 | 850 | XP_011540097.1 | |
| XM_017001812.1 | 4 | 1457 | XP_016857301.1 | |
| XM_017001813.1 | 4 | 1457 | XP_016857302.1 | |
| XM_017001814.1 | XP_016857303.1 | |||
| XM_017001815.1 | XP_016857304.1 | |||
| XM_017001816.1 | XP_016857305.1 | |||
| XM_017001817.1 | 6 | 775 | XP_016857306.1 | |
| XM_017001818.1 | 7 | 673 | XP_016857307.1 | |
| XM_017001819.1 | XP_016857308.1 | |||
| XM_017001820.1 | 7 | 673 | XP_016857309.1 | |
| XM_017001821.1 | XP_016857310.1 | |||
| XM_017001822.1 | XP_016857311.1 | |||
| XM_017001823.1 | 7 | 673 | XP_016857312.1 | |
| XM_017001824.1 | XP_016857313.1 | |||
| XM_017001825.1 | XP_016857314.1 | |||
| XM_017001826.1 | 5 | 1146 | XP_016857315.1 | |
| XM_017001827.1 | 6 | 1044 | XP_016857316.1 | |
| XM_017001828.1 | XP_016857317.1 | |||
| XM_017001829.1 | XP_016857318.1 | |||
| XM_017001830.1 | XP_016857319.1 | |||
| AF258548.1 | AAG23751.1 | |||
| AK075260.1 | 2 | 1257 | BAC11504.1 | |
| AK295232.1 | 7 | 868 | ||
| AL512700.1 | ||||
| AY358586.1 | AAQ88949.1 | |||
| BC004885.2 | AAH04885.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv300 | Chr.1:32206912 - 32241689 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map