Assay Details
Target Gene Details
Entrez Gene ID: | 80313 |
Gene Name: | leucine rich repeat containing 27 |
Gene Aliases: |
- |
Location: |
Chr.10:132330063-132381506 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC27 | NM_001143757.1 | 5 | 816 | NP_001137229.1 |
| NM_001143758.1 | 5 | 663 | NP_001137230.1 | |
| NM_001143759.1 | 5 | 663 | NP_001137231.1 | |
| NM_001309474.1 | 4 | 573 | NP_001296403.1 | |
| NM_030626.2 | 5 | 663 | NP_085129.1 | |
| XM_006717983.2 | 5 | 976 | XP_006718046.1 | |
| XM_006717984.2 | 5 | 663 | XP_006718047.1 | |
| XM_011540208.1 | 5 | 746 | XP_011538510.1 | |
| XM_011540209.1 | 5 | 663 | XP_011538511.1 | |
| XM_011540210.2 | 4 | 571 | XP_011538512.1 | |
| XM_017016690.1 | 5 | 663 | XP_016872179.1 | |
| XM_017016691.1 | 5 | 663 | XP_016872180.1 | |
| XM_017016692.1 | 5 | 663 | XP_016872181.1 | |
| XM_017016693.1 | 5 | 663 | XP_016872182.1 | |
| XM_017016694.1 | 4 | 550 | XP_016872183.1 | |
| AB051461.1 | 5 | 573 | ||
| AK093250.1 | 5 | 429 | BAC04108.1 | |
| AK097418.1 | 5 | 774 | ||
| AK098652.1 | 5 | 619 | BAC05366.1 | |
| AK295953.1 | 4 | 521 | ||
| AK301422.1 | 4 | 340 | ||
| BC050097.1 | 5 | 618 | AAH50097.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054569 | Chr.10:132009350 - 132351508 on Build GRCh38 | Gain |
|
| nsv1051555 | Chr.10:132295188 - 132415461 on Build GRCh38 | Gain |
|
| esv3624988 | Chr.10:132338306 - 132366249 on Build GRCh38 | Gain |
|
| nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
|
| esv2761642 | Chr.10:132048518 - 132365985 on Build GRCh38 | Gain |
|
| esv3624986 | Chr.10:132322882 - 132551350 on Build GRCh38 | Gain |
|
| nsv1052405 | Chr.10:132336481 - 132390781 on Build GRCh38 | Gain |
|
| esv3577872 | Chr.10:132342397 - 132412909 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73385054] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map