Assay Details
Target Gene Details
Entrez Gene ID: | 51304 |
Gene Name: | zinc finger DHHC-type containing 3 |
Gene Aliases: |
DHHC-3, GODZ, ZNF373 |
Location: |
Chr.3:44915261-44976190 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZDHHC3 | NM_001135179.1 | 7 | 1773 | NP_001128651.1 |
| NM_016598.2 | 8 | 1857 | NP_057682.1 | |
| XM_005265209.1 | XP_005265266.1 | |||
| XM_005265212.2 | XP_005265269.1 | |||
| XM_006713189.1 | XP_006713252.1 | |||
| XM_006713192.2 | 9 | 1959 | XP_006713255.1 | |
| XM_006713193.2 | 8 | 1876 | XP_006713256.1 | |
| XM_006713194.3 | XP_006713257.1 | |||
| XM_011533797.1 | XP_011532099.1 | |||
| XM_011533800.2 | XP_011532102.1 | |||
| XM_011533801.2 | 7 | 1744 | XP_011532103.1 | |
| XM_017006557.1 | XP_016862046.1 | |||
| XM_017006558.1 | XP_016862047.1 | |||
| XM_017006559.1 | XP_016862048.1 | |||
| XM_017006560.1 | XP_016862049.1 | |||
| XM_017006561.1 | XP_016862050.1 | |||
| XM_017006562.1 | XP_016862051.1 | |||
| XM_017006563.1 | XP_016862052.1 | |||
| XM_017006564.1 | XP_016862053.1 | |||
| XM_017006565.1 | XP_016862054.1 | |||
| XM_017006566.1 | 8 | 1811 | XP_016862055.1 | |
| XM_017006567.1 | 8 | 1849 | XP_016862056.1 | |
| XM_017006568.1 | XP_016862057.1 | |||
| XM_017006569.1 | 8 | 1848 | XP_016862058.1 | |
| XM_017006570.1 | XP_016862059.1 | |||
| XM_017006571.1 | 8 | 1853 | XP_016862060.1 | |
| AF052182.1 | 1 | 385 | ||
| AF247703.1 | 8 | 1797 | AAF63570.1 | |
| AK299782.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834675 | Chr.3:44805220 - 44966381 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map