Assay Details
Target Gene Details
Entrez Gene ID: | 266977 |
Gene Name: | adhesion G protein-coupled receptor F1 |
Gene Aliases: |
GPR110, KPG_012, PGR19, hGPCR36 |
Location: |
Chr.6:46997703-47042363 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRF1 | NM_153840.3 | NP_722582.2 | ||
| XM_011514469.1 | XP_011512771.1 | |||
| XM_017010735.1 | XP_016866224.1 | |||
| XM_017010736.1 | XP_016866225.1 | |||
| AB045123.1 | BAD83599.1 | |||
| AY140952.1 | AAN46666.1 | |||
| BU675866.1 | ||||
| CR627234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map