Assay Details
Target Gene Details
Entrez Gene ID: | 23429 |
Gene Name: | RING1 and YY1 binding protein |
Gene Aliases: |
AAP1, APAP-1, DEDAF, YEAF1 |
Location: |
Chr.3:72374593-72446623 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RYBP | NM_012234.6 | 4 | 1223 | NP_036366.3 |
| AB029551.1 | 4 | 1232 | BAA89486.1 | |
| AL049940.1 | 1 | 253 | ||
| BC036459.1 | 4 | 1228 | AAH36459.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954886 | Chr.3:72374650 - 72446149 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map