Assay Details
Target Gene Details
Entrez Gene ID: | 160364 |
Gene Name: | C-type lectin domain family 12 member A |
Gene Aliases: |
CD371, CLL-1, CLL1, DCAL-2, MICL |
Location: |
Chr.12:9951316-9995209 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC12A | NM_001207010.1 | NP_001193939.1 | ||
| NM_138337.5 | NP_612210.4 | |||
| NM_201623.3 | NP_963917.2 | |||
| XM_005253324.2 | XP_005253381.1 | |||
| XM_006719033.3 | XP_006719096.1 | |||
| XM_006719035.3 | XP_006719098.1 | |||
| XM_011520570.1 | XP_011518872.1 | |||
| XM_011520571.2 | XP_011518873.2 | |||
| XM_011520573.2 | XP_011518875.1 | |||
| AF247788.1 | AAL95693.1 | |||
| AK314001.1 | ||||
| AY426759.1 | AAR84594.1 | |||
| AY498550.1 | AAS00605.1 | |||
| AY498551.1 | AAS00606.1 | |||
| AY498552.1 | AAS00607.1 | |||
| AY547296.1 | AAT11783.1 | |||
| BC126289.1 | ||||
| BC126291.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1374n100 | Chr.12:9968689 - 10026775 on Build GRCh38 | Gain |
|
| nsv525593 | Chr.12:9968748 - 10004047 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map