Assay Details
Target Gene Details
Entrez Gene ID: | 4887 |
Gene Name: | neuropeptide Y receptor Y2 |
Gene Aliases: |
NPY2-R |
Location: |
Chr.4:155173514-155217078 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPY2R | NM_000910.3 | 1 | 62 | NP_000901.1 |
| XM_005263033.4 | XP_005263090.1 | |||
| XM_005263034.4 | 1 | 492 | XP_005263091.1 | |
| U42766.1 | 1 | 68 | AAB04120.1 |
More Information
Set Membership: |
|

Genomic Map