Assay Details
Target Gene Details
Entrez Gene ID: | 404220 |
Gene Name: | chromosome 6 open reading frame 201 |
Gene Aliases: |
dJ1013A10.5 |
Location: |
Chr.6:4079197-4130788 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C6orf201 | NM_001085401.2 | 5 | 1189 | NP_001078870.1 |
| NR_104463.1 | 7 | 2606 | ||
| NR_104464.1 | 5 | 1952 | ||
| XM_017010861.1 | 4 | 820 | XP_016866350.1 | |
| AF429972.1 | 4 | 639 | AAP97304.1 | |
| AK124114.1 | 6 | 2041 | ||
| BC047663.1 | 5 | 1186 | AAH47663.1 |
Target Gene Details
Entrez Gene ID: | 10455 |
Gene Name: | enoyl-CoA delta isomerase 2 |
Gene Aliases: |
ACBD2, DRS-1, DRS1, HCA88, PECI, dJ1013A10.3 |
Location: |
Chr.6:4115693-4135597 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECI2 | NM_001166010.1 | NP_001159482.1 | ||
| NM_006117.2 | NP_006108.2 | |||
| NM_206836.2 | NP_996667.2 | |||
| NR_028588.1 | ||||
| XM_006714957.1 | XP_006715020.1 | |||
| XM_017010170.1 | XP_016865659.1 | |||
| AB209917.1 | BAD93154.1 | |||
| AF069301.1 | AAC19317.1 | |||
| AF153612.1 | AAD34173.1 | |||
| AF244138.1 | AAF66247.1 | |||
| AF257175.1 | AAF68974.1 | |||
| AK057175.1 | ||||
| AK075108.1 | ||||
| AK222617.1 | BAD96337.1 | |||
| AK223003.1 | BAD96723.1 | |||
| AK297049.1 | ||||
| AL136642.1 | CAB66577.1 | |||
| AY297537.1 | ||||
| BC002668.2 | AAH02668.3 | |||
| BC016781.1 | AAH16781.1 | |||
| BC017474.1 | AAH17474.1 | |||
| BC033841.1 | AAH33841.3 | |||
| BC034702.1 | AAH34702.1 | |||
| CR456768.1 | CAG33049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422457 | Chr.6:4100099 - 4377162 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map