Assay Details
Target Gene Details
Entrez Gene ID: | 101928461 |
Gene Name: | uncharacterized LOC101928461 |
Gene Aliases: |
- |
Location: |
Chr.6:136629114-136648198 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928461 | NR_125858.1 | |||
| AI138300.1 | ||||
| BX093515.1 | ||||
| BX398118.2 |
Target Gene Details
Entrez Gene ID: | 4217 |
Gene Name: | mitogen-activated protein kinase kinase kinase 5 |
Gene Aliases: |
ASK1, MAPKKK5, MEKK5 |
Location: |
Chr.6:136557046-136793098 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K5 | NM_005923.3 | NP_005914.1 | ||
| XM_011535839.2 | XP_011534141.2 | |||
| XM_017010870.1 | XP_016866359.1 | |||
| XM_017010871.1 | XP_016866360.1 | |||
| XM_017010872.1 | XP_016866361.1 | |||
| XM_017010873.1 | XP_016866362.1 | |||
| XM_017010874.1 | XP_016866363.1 | |||
| XM_017010875.1 | XP_016866364.1 | |||
| XM_017010876.1 | XP_016866365.1 | |||
| XM_017010877.1 | XP_016866366.1 | |||
| AB209093.1 | BAD92330.1 | |||
| AK293953.1 | ||||
| AK294507.1 | ||||
| BC054503.2 | AAH54503.1 | |||
| BC088829.1 | AAH88829.1 | |||
| D84476.2 | BAA12684.2 | |||
| U67156.1 | AAC50894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464064 | Chr.6:136476073 - 136676481 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map