Assay Details
Target Gene Details
Entrez Gene ID: | 892 |
Gene Name: | cyclin C |
Gene Aliases: |
CycC |
Location: |
Chr.6:99542387-99568973 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNC | NM_001013399.1 | NP_001013417.1 | ||
| NM_005190.3 | NP_005181.2 | |||
| XM_006715594.1 | XP_006715657.1 | |||
| AB125137.1 | BAD00144.1 | |||
| AK222768.1 | ||||
| AK223247.1 | ||||
| AK291914.1 | ||||
| AK298558.1 | ||||
| BC010135.2 | AAH10135.1 | |||
| BC041123.1 | ||||
| BC056153.1 | AAH56153.1 | |||
| M74091.1 |
Target Gene Details
Entrez Gene ID: | 100130890 |
Gene Name: | thiosulfate sulfurtransferase (rhodanese)-like domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.6:99424854-99579288 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSTD3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1029536 | Chr.6:98422726 - 99818247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map