Assay Details
Target Gene Details
Entrez Gene ID: | 64794 |
Gene Name: | DEAD-box helicase 31 |
Gene Aliases: |
PPP1R25 |
Location: |
Chr.9:132592997-132670403 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX31 | NM_001322340.1 | 18 | 2838 | NP_001309269.1 |
| NM_001322341.1 | 21 | 3161 | NP_001309270.1 | |
| NM_001322342.1 | 20 | 2561 | NP_001309271.1 | |
| NM_001322343.1 | 20 | 2620 | NP_001309272.1 | |
| NM_022779.8 | 20 | 3057 | NP_073616.6 | |
| NR_136309.1 | 19 | 2920 | ||
| XM_005272207.4 | 20 | 2744 | XP_005272264.1 | |
| XM_006717236.3 | 18 | 2865 | XP_006717299.1 | |
| AF335568.1 | 20 | 3058 | AAQ14889.1 | |
| AF335569.1 | 18 | 2839 | AAQ14890.1 | |
| AF427339.1 | 20 | 3058 | AAL26549.1 | |
| AK023695.1 | 16 | 2130 | BAB14644.1 | |
| AK027002.1 | 20 | 2641 | BAB15620.1 | |
| AK225121.1 | 20 | 2612 | ||
| AK225350.1 | 20 | 2642 | ||
| BC012726.2 | 20 | 2683 | AAH12726.2 |
More Information
Additional Information:
For this assay, SNP(s) [rs73560850] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map