Assay Details
Target Gene Details
Entrez Gene ID: | 55835 |
Gene Name: | centromere protein J |
Gene Aliases: |
BM032, CENP-J, CPAP, LAP, LIP1, MCPH6, SASS4, SCKL4, Sas-4 |
Location: |
Chr.13:24882274-24922889 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPJ | NM_018451.4 | NP_060921.3 | ||
| NR_047594.1 | ||||
| NR_047595.1 | ||||
| XM_011535149.2 | XP_011533451.1 | |||
| XM_011535150.2 | XP_011533452.1 | |||
| XM_017020673.1 | XP_016876162.1 | |||
| AF139625.1 | AAG21074.1 | |||
| AK292406.1 | ||||
| AL833182.1 | CAI46195.1 | |||
| BC113110.1 | AAI13111.1 | |||
| BC113111.1 | AAI13112.1 | |||
| BC113662.1 | ||||
| BC113664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041986 | Chr.13:24870991 - 24986494 on Build GRCh38 | Gain |
|
| nsv1053023 | Chr.13:24866993 - 24916871 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map