Assay Details
Target Gene Details
Entrez Gene ID: | 118490 |
Gene Name: | MSS51 mitochondrial translational activator |
Gene Aliases: |
ZMYND17 |
Location: |
Chr.10:73423579-73433561 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSS51 | NM_001024593.1 | 7 | 1495 | NP_001019764.1 |
| AK096884.1 | 7 | 1065 | BAC04883.1 | |
| AK309870.1 | 8 | 1551 | ||
| BC094693.1 | 7 | 1474 | AAH94693.1 | |
| BC109255.1 | 6 | 1408 | AAI09256.1 | |
| BC109256.1 | 6 | 1408 | AAI09257.1 | |
| BX537678.1 | 5 | 2620 | CAD97808.1 | |
| CN395159.1 | 2 | 336 | ||
| CR936824.1 | 6 | 1802 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831911 | Chr.10:73313880 - 73483446 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map