Assay Details
Target Gene Details
Entrez Gene ID: | 4668 |
Gene Name: | alpha-N-acetylgalactosaminidase |
Gene Aliases: |
D22S674, GALB |
Location: |
Chr.22:42058334-42070870 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAGA | NM_000262.2 | 4 | 974 | NP_000253.1 |
| XM_005261615.4 | 5 | 613 | XP_005261672.1 | |
| XM_005261616.4 | 5 | 595 | XP_005261673.1 | |
| BC000095.2 | 4 | 510 | AAH00095.1 | |
| CR456527.1 | 4 | 483 | CAG30413.1 | |
| M29276.1 | 4 | 772 | AAA59902.1 | |
| M38083.1 | 4 | 501 | AAA36351.1 | |
| M62783.1 | 4 | 898 | AAA51677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508737 | Chr.22:42052007 - 42134336 on Build GRCh38 | Insertion |
|
| nsv834210 | Chr.22:42057329 - 42209100 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map