Assay Details
Target Gene Details
Entrez Gene ID: | 23780 |
Gene Name: | apolipoprotein L2 |
Gene Aliases: |
APOL-II, APOL3 |
Location: |
Chr.22:36226209-36239954 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOL2 | NM_030882.3 | 5 | 933 | NP_112092.2 |
| NM_145637.2 | 6 | 1074 | NP_663612.2 | |
| XM_011530074.1 | 6 | 1640 | XP_011528376.1 | |
| XM_011530075.1 | 7 | 1646 | XP_011528377.1 | |
| XM_011530076.2 | 6 | 951 | XP_011528378.1 | |
| XM_011530077.2 | 5 | 1029 | XP_011528379.1 | |
| XM_011530078.1 | 7 | 1740 | XP_011528380.1 | |
| XM_017028724.1 | 6 | 948 | XP_016884213.1 | |
| AB208990.1 | 5 | 833 | BAD92227.1 | |
| AF305225.1 | 5 | 817 | AAK20211.1 | |
| AF305429.1 | 5 | 796 | AAL09359.1 | |
| AF324223.1 | 5 | 933 | AAK11592.1 | |
| AF324224.1 | 6 | 1074 | AAK11593.1 | |
| AK056938.1 | 5 | 865 | BAB71315.1 | |
| AK303978.1 | 6 | 942 | ||
| BC004395.1 | 5 | 801 | AAH04395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647680 | Chr.22:36200214 - 36234114 on Build GRCh38 | Loss |
|
| esv3647682 | Chr.22:36224551 - 36308255 on Build GRCh38 | Gain |
|
| nsv522931 | Chr.22:36220399 - 36318590 on Build GRCh38 | Gain |
|
| esv1003985 | Chr.22:36227668 - 36241537 on Build GRCh38 | Deletion |
|
| dgv4564n100 | Chr.22:36218252 - 36319792 on Build GRCh38 | Gain |
|
| esv2760715 | Chr.22:36218264 - 36317486 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map