Assay Details
Target Gene Details
Entrez Gene ID: | 9276 |
Gene Name: | coatomer protein complex subunit beta 2 |
Gene Aliases: |
beta'-COP |
Location: |
Chr.3:139355595-139389680 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPB2 | NM_004766.2 | 22 | 3000 | NP_004757.1 |
| NR_023350.1 | 22 | 3079 | ||
| XM_017007512.1 | 23 | 3185 | XP_016863001.1 | |
| XM_017007513.1 | 21 | 2746 | XP_016863002.1 | |
| AK302943.1 | 22 | 2913 | ||
| BC000326.2 | 22 | 2946 | AAH00326.1 | |
| X70476.1 | 22 | 2938 | CAA49900.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4034 | Chr.3:139316134 - 139361291 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map