Assay Details
Target Gene Details
Entrez Gene ID: | 30 |
Gene Name: | acetyl-CoA acyltransferase 1 |
Gene Aliases: |
ACAA, PTHIO, THIO |
Location: |
Chr.3:38122710-38137242 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAA1 | NM_001130410.1 | NP_001123882.1 | ||
| NM_001607.3 | NP_001598.1 | |||
| NR_024024.1 | ||||
| XM_006713122.1 | XP_006713185.1 | |||
| XM_006713123.2 | XP_006713186.1 | |||
| XM_011533650.2 | XP_011531952.1 | |||
| AF035295.1 | AAB88181.1 | |||
| AK025138.1 | ||||
| AK127051.1 | 6 | 4330 | ||
| AK293716.1 | ||||
| BC000635.2 | AAH00635.1 | |||
| BC011977.1 | AAH11977.1 | |||
| BC014474.1 | AAH14474.1 | |||
| X12966.1 | CAA31412.1 | |||
| X14813.1 | CAA32918.1 | |||
| X51460.1 | CAA35825.1 |
More Information
Set Membership: |
|

Genomic Map