Assay Details
Target Gene Details
Entrez Gene ID: | 55171 |
Gene Name: | TBCC domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.3:186546067-186570543 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBCCD1 | NM_001134415.1 | 6 | 1872 | NP_001127887.1 |
| NM_001286749.1 | 5 | 1493 | NP_001273678.1 | |
| NM_018138.4 | 6 | 1523 | NP_060608.1 | |
| AK001422.1 | 6 | 1496 | BAA91682.1 | |
| AK124362.1 | 5 | 1268 | ||
| BC025748.1 | 6 | 1509 | AAH25748.1 |
More Information
Set Membership: |
|

Genomic Map