Assay Details
Target Gene Details
Entrez Gene ID: | 8751 |
Gene Name: | ADAM metallopeptidase domain 15 |
Gene Aliases: |
MDC15 |
Location: |
Chr.1:155051272-155062776 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM15 | NM_001261464.1 | 21 | 2659 | NP_001248393.1 |
| NM_001261465.1 | 21 | 2631 | NP_001248394.1 | |
| NM_003815.4 | 21 | 2629 | NP_003806.3 | |
| NM_207191.2 | 20 | 2559 | NP_997074.1 | |
| NM_207194.2 | 22 | 2704 | NP_997077.1 | |
| NM_207195.2 | 22 | 2701 | NP_997078.1 | |
| NM_207196.2 | 23 | 2773 | NP_997079.1 | |
| NM_207197.2 | 23 | 2776 | NP_997080.1 | |
| NR_048577.1 | 23 | 2915 | ||
| NR_048578.1 | 23 | 2918 | ||
| NR_048579.1 | 22 | 2843 | ||
| AB209157.1 | 22 | 4304 | BAD92394.1 | |
| AK000667.1 | 13 | 1325 | ||
| AK075498.1 | 13 | 1703 | ||
| AK296925.1 | 21 | 2580 | ||
| AY560593.1 | 20 | 2545 | AAS72991.1 | |
| AY560594.1 | 21 | 2615 | AAS72992.1 | |
| AY560595.1 | 21 | 2617 | AAS72993.1 | |
| AY560596.1 | 22 | 2690 | AAS72994.1 | |
| AY560597.1 | 22 | 2687 | AAS72995.1 | |
| AY560598.1 | 23 | 2759 | AAS72996.1 | |
| AY560599.1 | 23 | 2762 | AAS72997.1 | |
| AY560600.1 | 23 | 2901 | AAS72998.1 | |
| AY560601.1 | 23 | 2904 | AAS72999.1 | |
| AY576417.1 | 22 | 2829 | AAS73000.1 | |
| BC014566.2 | 21 | 2595 | AAH14566.1 | |
| BM971880.1 | 1 | 224 | ||
| U41767.1 | 21 | 2521 | AAC50404.1 | |
| U46005.1 | 21 | 2514 | AAC51112.1 |
Target Gene Details
Entrez Gene ID: | 100505666 |
Gene Name: | uncharacterized LOC100505666 |
Gene Aliases: |
- |
Location: |
Chr.1:155045191-155063991 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100505666 | NR_040773.1 | |||
| BM047608.1 | ||||
| DB152307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| esv3587616 | Chr.1:155055926 - 155098657 on Build GRCh38 | Gain |
|
| esv3587615 | Chr.1:155047853 - 155175013 on Build GRCh38 | Gain |
|
| nsv952212 | Chr.1:154998225 - 155087624 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map