Assay Details
Target Gene Details
Entrez Gene ID: | 221613 |
Gene Name: | histone cluster 1, H2aa |
Gene Aliases: |
H2AA, H2AFR, TH2A, bA317E16.2 |
Location: |
Chr.6:25726063-25726562 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST1H2AA | NM_170745.3 | 1 | 189 | NP_734466.1 |
| BC062211.1 | 1 | 193 | AAH62211.1 |
Target Gene Details
Entrez Gene ID: | 6568 |
Gene Name: | solute carrier family 17 member 1 |
Gene Aliases: |
NAPI-1, NPT-1, NPT1 |
Location: |
Chr.6:25723744-25832108 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A1 | XM_017011201.1 | XP_016866690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3570813 | Chr.6:25724901 - 25728340 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map