Assay Details
Target Gene Details
Entrez Gene ID: | 10007 |
Gene Name: | glucosamine-6-phosphate deaminase 1 |
Gene Aliases: |
GNP1, GNPDA, GNPI, GPI, HLN |
Location: |
Chr.5:142000669-142013055 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNPDA1 | NM_005471.4 | 7 | 1046 | NP_005462.1 |
| XM_005268348.1 | 7 | 1303 | XP_005268405.1 | |
| XM_006714747.2 | 8 | 1070 | XP_006714810.1 | |
| XM_011537537.1 | 8 | 1365 | XP_011535839.1 | |
| AF029914.1 | 6 | 992 | AAB84217.1 | |
| AJ002231.1 | 6 | 1738 | CAA05259.1 | |
| AK022779.1 | 6 | 916 | ||
| AK290686.1 | 6 | 1000 | ||
| AK296452.1 | 6 | 787 | ||
| BC012853.2 | 6 | 984 | AAH12853.1 | |
| BC020769.1 | 7 | 1018 | AAH20769.1 | |
| BC022322.1 | 7 | 1018 | AAH22322.1 | |
| D31766.1 | 8 | 1380 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1154058 | Chr.5:141632251 - 142199943 on Build GRCh38 | Deletion |
|
| nsv5036 | Chr.5:141978308 - 142008100 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map