Assay Details
Target Gene Details
Entrez Gene ID: | 55010 |
Gene Name: | PARP1 binding protein |
Gene Aliases: |
AROM, C12orf48, PARI |
Location: |
Chr.12:102120171-102197520 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARPBP | NM_001319988.1 | 12 | 3098 | NP_001306917.1 |
| NM_001319993.1 | 12 | 2975 | NP_001306922.1 | |
| NM_001319994.1 | 11 | 2862 | NP_001306923.1 | |
| NM_017915.4 | 11 | 2867 | NP_060385.3 | |
| NR_135123.1 | 5 | 1947 | ||
| XM_011538509.1 | 10 | 2633 | XP_011536811.1 | |
| XM_011538514.2 | 10 | 2680 | XP_011536816.1 | |
| XM_011538515.1 | 9 | 2473 | XP_011536817.1 | |
| XM_017019534.1 | 10 | 2696 | XP_016875023.1 | |
| XM_017019535.1 | 10 | 2706 | XP_016875024.1 | |
| XM_017019536.1 | 9 | 2609 | XP_016875025.1 | |
| XM_017019537.1 | 9 | 2504 | XP_016875026.1 | |
| XM_017019538.1 | 10 | 2731 | XP_016875027.1 | |
| XM_017019539.1 | 8 | 2270 | XP_016875028.1 | |
| XM_017019540.1 | 9 | 2466 | XP_016875029.1 | |
| XM_017019542.1 | 8 | 2473 | XP_016875031.1 | |
| XM_017019543.1 | 8 | 2362 | XP_016875032.1 | |
| XM_017019544.1 | 7 | 2283 | XP_016875033.1 | |
| AA757264.1 | 1 | 225 | ||
| AK000648.1 | BAA91306.1 | |||
| BC098313.1 | 8 | 2257 | AAH98313.1 | |
| BC099734.1 | 6 | 2071 | ||
| BC099844.1 | 7 | 2150 | ||
| CA438709.1 | 1 | 239 |
Target Gene Details
Entrez Gene ID: | 5367 |
Gene Name: | pro-melanin concentrating hormone |
Gene Aliases: |
MCH, ppMCH |
Location: |
Chr.12:102196459-102199540 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMCH | NM_002674.3 | NP_002665.2 | ||
| XM_017019483.1 | XP_016874972.1 | |||
| BC018048.1 | AAH18048.1 | |||
| CB047052.1 | ||||
| CB047053.1 | ||||
| M57703.1 | AAA63214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560033 | Chr.12:102163421 - 102233833 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map