Assay Details
Target Gene Details
Entrez Gene ID: | 79133 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 5 |
Gene Aliases: |
C20orf7, bA526K24.2, dJ842G6.1 |
Location: |
Chr.20:13785026-13821580 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF5 | NM_001039375.2 | 1 | 180 | NP_001034464.1 |
| NM_024120.4 | 1 | 180 | NP_077025.2 | |
| NR_029377.1 | 1 | 180 | ||
| XM_006723620.3 | 1 | 157 | XP_006723683.1 | |
| XM_006723622.2 | 1 | 165 | XP_006723685.1 | |
| XM_006723623.2 | 1 | 143 | XP_006723686.1 | |
| XM_006723624.2 | 1 | 177 | XP_006723687.1 | |
| XM_011529342.2 | 1 | 157 | XP_011527644.1 | |
| XM_017028061.1 | XP_016883550.1 | |||
| AK091060.1 | 1 | 157 | ||
| AK289781.1 | 1 | 172 | ||
| BC005984.1 | AAH05984.1 | |||
| BC073158.1 | 1 | 157 | AAH73158.1 | |
| CB156360.1 | 1 | 170 | ||
| DA289766.1 | 1 | 180 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061303 | Chr.20:13414899 - 13830344 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map