Assay Details
Target Gene Details
Entrez Gene ID: | 10659 |
Gene Name: | CUGBP, Elav-like family member 2 |
Gene Aliases: |
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR |
Location: |
Chr.10:10462515-11336675 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Target Gene Details
Entrez Gene ID: | 414196 |
Gene Name: | CELF2 antisense RNA 1 |
Gene Aliases: |
C10orf31 |
Location: |
Chr.10:11316833-11344674 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELF2-AS1 | NR_126062.1 | 3 | 595 | |
| AK097813.1 | 2 | 491 | BAC05176.1 |
More Information
Set Membership: |
|

Genomic Map