Assay Details
Target Gene Details
Entrez Gene ID: | 6770 |
Gene Name: | steroidogenic acute regulatory protein |
Gene Aliases: |
STARD1 |
Location: |
Chr.8:38141747-38151265 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAR | NM_000349.2 | 7 | 2421 | NP_000340.2 |
| XM_006716392.2 | 6 | 2510 | XP_006716455.1 | |
| AV706111.1 | 1 | 322 | ||
| AV707696.1 | 1 | 453 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616917 | Chr.8:38078635 - 38160673 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map