Assay Details
Target Gene Details
Entrez Gene ID: | 151126 |
Gene Name: | zinc finger protein 385B |
Gene Aliases: |
ZNF533 |
Location: |
Chr.2:179441982-179864550 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF385B | NM_001113397.1 | 8 | 1875 | NP_001106868.1 |
| NM_001113398.1 | 8 | 1735 | NP_001106869.1 | |
| NM_001282725.1 | 10 | 1829 | NP_001269654.1 | |
| NM_152520.4 | 10 | 2402 | NP_689733.3 | |
| NR_104234.1 | ||||
| XM_011510713.2 | 12 | 5543 | XP_011509015.1 | |
| XM_011510714.2 | 11 | 5493 | XP_011509016.1 | |
| XM_011510715.2 | 11 | 5446 | XP_011509017.1 | |
| XM_011510716.2 | 11 | 2654 | XP_011509018.1 | |
| XM_011510717.2 | 11 | 2651 | XP_011509019.1 | |
| XM_011510719.2 | 11 | 1914 | XP_011509021.1 | |
| XM_011510720.2 | 10 | 1774 | XP_011509022.1 | |
| XM_011510721.2 | 9 | 4067 | XP_011509023.1 | |
| XM_017003435.1 | 10 | 2083 | XP_016858924.1 | |
| XM_017003436.1 | 10 | 2561 | XP_016858925.1 | |
| XM_017003437.1 | 9 | 1718 | XP_016858926.1 | |
| XM_017003438.1 | 8 | 7201 | XP_016858927.1 | |
| XM_017003439.1 | 8 | 1709 | XP_016858928.1 | |
| XM_017003440.1 | 9 | 5238 | XP_016858929.1 | |
| AK057999.1 | 9 | 1618 | BAB71629.1 | |
| AK096822.1 | 8 | 1875 | BAC04870.1 | |
| BC038422.1 | 10 | 1827 | AAH38422.1 | |
| BC048123.1 | AAH48123.1 | |||
| BC092423.1 | 10 | 2237 | AAH92423.1 |
More Information
Set Membership: |
|

Genomic Map