Assay Details
Target Gene Details
Entrez Gene ID: | 8925 |
Gene Name: | HECT and RLD domain containing E3 ubiquitin protein ligase family member 1 |
Gene Aliases: |
p532, p619 |
Location: |
Chr.15:63608618-63833964 on Build GRCh38 |
Assay Gene Location: | Within Exon 80 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HERC1 | NM_003922.3 | 78 | 14722 | NP_003913.3 |
| XM_017022699.1 | 79 | 14879 | XP_016878188.1 | |
| XM_017022700.1 | 79 | 14843 | XP_016878189.1 | |
| XM_017022701.1 | 79 | 14828 | XP_016878190.1 | |
| XM_017022702.1 | 78 | 14828 | XP_016878191.1 | |
| XM_017022703.1 | 79 | 14825 | XP_016878192.1 | |
| XM_017022704.1 | 79 | 14807 | XP_016878193.1 | |
| XM_017022705.1 | 78 | 14801 | XP_016878194.1 | |
| AF131824.1 | 1 | 1017 | ||
| AK091624.1 | 1 | 1897 | ||
| AK226162.1 | 65 | 11785 | ||
| BC040929.1 | 12 | 1946 | AAH40929.1 | |
| U50078.1 | 78 | 14670 | AAD12586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833034 | Chr.15:63500671 - 63657512 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77087691] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map