Assay Details
Target Gene Details
Entrez Gene ID: | 54880 |
Gene Name: | BCL6 corepressor |
Gene Aliases: |
ANOP2, MAA2, MCOPS2 |
Location: |
Chr.X:40051246-40177390 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCOR | NM_001123383.1 | 15 | 5799 | NP_001116855.1 |
| NM_001123384.1 | 14 | 5745 | NP_001116856.1 | |
| NM_001123385.1 | 15 | 5830 | NP_001116857.1 | |
| NM_017745.5 | 15 | 5728 | NP_060215.4 | |
| XM_005272616.1 | 15 | 5962 | XP_005272673.1 | |
| XM_005272618.3 | 15 | 5628 | XP_005272675.1 | |
| XM_005272619.3 | 14 | 6234 | XP_005272676.1 | |
| XM_005272620.3 | 14 | 6132 | XP_005272677.1 | |
| XM_006724536.3 | 15 | 7064 | XP_006724599.1 | |
| XM_011543929.2 | 15 | 5738 | XP_011542231.1 | |
| XM_011543930.1 | 15 | 5751 | XP_011542232.1 | |
| XM_011543931.2 | 15 | 5861 | XP_011542233.1 | |
| XM_017029615.1 | 15 | 6962 | XP_016885104.1 | |
| XM_017029616.1 | 15 | 6336 | XP_016885105.1 | |
| AF317391.1 | AAG41429.1 | |||
| AK000292.1 | 7 | 1605 | BAA91061.1 | |
| AK074286.1 | 11 | 4736 | BAB85037.1 | |
| AK095360.1 | 10 | 2439 | ||
| AK225581.1 | 7 | 1558 | ||
| AY316592.1 | AAR08265.1 | |||
| BC009675.2 | 6 | 1356 | AAH09675.2 | |
| BC063536.1 | 4 | 912 | AAH63536.1 | |
| BC114220.1 | 14 | 5745 | AAI14221.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs35289701] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map