Assay Details
Target Gene Details
Entrez Gene ID: | 54888 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 2 |
Gene Aliases: |
MISU, MRT5, SAKI, TRM4 |
Location: |
Chr.5:6599239-6633360 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSUN2 | NM_001193455.1 | 18 | 2457 | NP_001180384.1 |
| NM_017755.5 | 19 | 2562 | NP_060225.4 | |
| NR_037947.1 | 18 | 2477 | ||
| AK000310.1 | 13 | 1558 | BAA91075.1 | |
| AK023994.1 | 7 | 828 | BAB14762.1 | |
| AK055456.1 | 18 | 2407 | ||
| AK291144.1 | 19 | 2245 | ||
| AK298980.1 | 18 | 2139 | ||
| BC001041.2 | 17 | 1918 | AAH01041.3 | |
| BC137083.1 | 19 | 2243 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894117 | Chr.5:6511201 - 6616164 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map