Assay Details
Target Gene Details
Entrez Gene ID: | 24140 |
Gene Name: | FtsJ RNA methyltransferase homolog 1 (E. coli) |
Gene Aliases: |
CDLIV, JM23, MRX44, MRX9, SPB1, TRMT7 |
Location: |
Chr.X:48476021-48486364 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FTSJ1 | NM_001282157.1 | NP_001269086.1 | ||
| NM_012280.3 | NP_036412.1 | |||
| NM_177439.2 | NP_803188.1 | |||
| XM_005272595.1 | XP_005272652.1 | |||
| XM_011543893.2 | XP_011542195.1 | |||
| XM_011543894.2 | XP_011542196.1 | |||
| XM_011543895.1 | XP_011542197.1 | |||
| XM_017029382.1 | XP_016884871.1 | |||
| AF063015.1 | AAC33734.1 | |||
| AJ005892.1 | CAA06749.1 | |||
| AK024023.1 | ||||
| AK297486.1 | ||||
| AK315138.1 | ||||
| BC023584.1 | AAH23584.1 | |||
| KU178580.1 | ||||
| KU178581.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
| esv3573946 | Chr.X:48471017 - 48479788 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map