Assay Details
Target Gene Details
Entrez Gene ID: | 9874 |
Gene Name: | tousled like kinase 1 |
Gene Aliases: |
PKU-beta |
Location: |
Chr.2:170990823-171231314 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLK1 | NM_001136554.1 | 21 | 2580 | NP_001130026.1 |
| NM_001136555.1 | 19 | 2351 | NP_001130027.1 | |
| NM_012290.4 | 21 | 3021 | NP_036422.3 | |
| XM_011512238.2 | 20 | 2872 | XP_011510540.1 | |
| XM_017005421.1 | 22 | 3130 | XP_016860910.1 | |
| XM_017005422.1 | 20 | 2942 | XP_016860911.1 | |
| AB004885.1 | 22 | 2824 | BAA20562.1 | |
| AF162666.1 | AAF03094.1 | |||
| AK090779.1 | ||||
| BC032657.1 | 21 | 2582 | AAH32657.1 | |
| D50927.2 | 21 | 2654 |
More Information
Set Membership: |
|

Genomic Map