Assay Details
Target Gene Details
Entrez Gene ID: | 195814 |
Gene Name: | short chain dehydrogenase/reductase family 16C, member 5 |
Gene Aliases: |
EPHD-2, RDH#2, RDH-E2, RDHE2, retSDR2 |
Location: |
Chr.8:56299648-56320776 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDR16C5 | NM_001318049.1 | 8 | 2478 | NP_001304978.1 |
| NM_001318050.1 | 6 | 2199 | NP_001304979.1 | |
| NM_138969.3 | 7 | 2331 | NP_620419.2 | |
| XM_011517479.1 | 7 | 2346 | XP_011515781.1 | |
| AK057667.1 | 7 | 1674 | BAB71545.1 | |
| AK294634.1 | 6 | 1602 | ||
| AY444559.1 | 6 | 1598 | AAS68535.1 | |
| BC037219.1 | 7 | 1678 | AAH37219.2 | |
| BC064525.1 | 7 | 1650 | AAH64525.1 |
More Information
Set Membership: |
|

Genomic Map