Assay Details
Target Gene Details
Entrez Gene ID: | 7068 |
Gene Name: | thyroid hormone receptor beta |
Gene Aliases: |
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2 |
Location: |
Chr.3:24117153-24495281 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THRB | NM_000461.4 | 10 | 6100 | NP_000452.2 |
| NM_001128176.2 | 11 | 6211 | NP_001121648.1 | |
| NM_001128177.1 | 11 | 6237 | NP_001121649.1 | |
| NM_001252634.1 | 12 | 6335 | NP_001239563.1 | |
| XM_006713317.3 | 11 | 6451 | XP_006713380.1 | |
| AK096885.1 | ||||
| AK098084.1 | 1 | 2400 | ||
| GQ869477.1 | 1 | 4429 | ||
| GQ869478.1 | 2 | 4306 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834639 | Chr.3:23995622 - 24192434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map