Assay Details
Target Gene Details
Entrez Gene ID: | 9368 |
Gene Name: | SLC9A3 regulator 1 |
Gene Aliases: |
EBP50, NHERF, NHERF-1, NHERF1, NPHLOP2 |
Location: |
Chr.17:74748612-74769360 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A3R1 | NM_004252.4 | NP_004243.1 | ||
| AF015926.1 | AAC52084.1 | |||
| AF036241.1 | AAC04572.1 | |||
| AK026581.1 | ||||
| AK094467.1 | ||||
| AK128474.1 | ||||
| AK312498.1 | ||||
| BC001443.2 | AAH01443.1 | |||
| BC003361.1 | AAH03361.1 | |||
| BC011777.1 | AAH11777.1 | |||
| BC049220.1 | AAH49220.1 | |||
| BC053350.1 | AAH53350.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
| nsv523611 | Chr.17:74755524 - 74767659 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map