Assay Details
Target Gene Details
Entrez Gene ID: | 51227 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class P |
Gene Aliases: |
DCRC, DCRC-S, DSCR5, DSRC, PIG-P |
Location: |
Chr.21:37065364-37073158 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGP | NM_001320480.1 | NP_001307409.1 | ||
| NM_016430.3 | NP_057514.2 | |||
| NM_153681.2 | NP_710148.1 | |||
| NM_153682.2 | NP_710149.1 | |||
| XM_005260990.4 | XP_005261047.1 | |||
| XM_011529596.2 | XP_011527898.1 | |||
| XM_017028364.1 | XP_016883853.1 | |||
| XM_017028365.1 | XP_016883854.1 | |||
| AB035742.1 | BAA95633.1 | |||
| AB035743.1 | BAA95634.1 | |||
| AB035744.1 | BAA95635.1 | |||
| AB035745.1 | BAA95636.1 | |||
| AB037162.1 | BAA96871.1 | |||
| AB037163.1 | BAA96872.1 | |||
| AB037164.1 | BAA96873.1 | |||
| AB039659.1 | BAB12395.1 | |||
| AB451328.1 | ||||
| AB451472.1 | ||||
| AF216305.1 | AAF32289.1 | |||
| AF237812.1 | AAG09757.1 | |||
| AI127387.1 | ||||
| AK026311.1 | ||||
| AK304656.1 | ||||
| AK314457.1 | ||||
| AK316609.1 | ||||
| BC005180.1 | AAH05180.1 | |||
| BC011007.1 | AAH11007.1 | |||
| BE275469.1 | ||||
| BT007053.1 | AAP35702.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575389 | Chr.21:37022665 - 37114044 on Build GRCh38 | Gain |
|
| nsv3510 | Chr.21:37054434 - 37086086 on Build GRCh38 | Insertion |
|
| nsv1066107 | Chr.21:37045398 - 37132761 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map