Assay Details
Target Gene Details
Entrez Gene ID: | 27242 |
Gene Name: | TNF receptor superfamily member 21 |
Gene Aliases: |
BM-018, CD358, DR6 |
Location: |
Chr.6:47231527-47309976 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF21 | NM_014452.4 | NP_055267.1 | ||
| AB209394.1 | BAD92631.1 | |||
| AF068868.1 | AAC34583.1 | |||
| AJ420531.1 | ||||
| AK001504.1 | ||||
| AK315560.1 | ||||
| AY358304.1 | AAQ88671.1 | |||
| BC010241.1 | AAH10241.1 | |||
| BC017730.2 | AAH17730.1 | |||
| BC021572.1 | ||||
| BT007420.1 | AAP36088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map