Assay Details
Target Gene Details
Entrez Gene ID: | 1009 |
Gene Name: | cadherin 11 |
Gene Aliases: |
CAD11, CDHOB, OB, OSF-4 |
Location: |
Chr.16:64943753-65126112 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH11 | NM_001308392.1 | 14 | 3065 | NP_001295321.1 |
| NM_001797.3 | 13 | 2886 | NP_001788.2 | |
| XM_005255761.3 | 13 | 2757 | XP_005255818.1 | |
| XM_005255762.2 | 13 | 2971 | XP_005255819.1 | |
| XM_005255763.2 | 13 | 2681 | XP_005255820.1 | |
| XM_011522803.2 | 12 | 2591 | XP_011521105.1 | |
| XM_017022830.1 | 12 | 2446 | XP_016878319.1 | |
| AB209760.1 | 11 | 2364 | BAD92997.1 | |
| AK291251.1 | 13 | 2812 | ||
| AK297377.1 | 13 | 2397 | ||
| BC013609.1 | 11 | 2505 | AAH13609.1 | |
| D21254.1 | 13 | 2772 | BAA04798.1 | |
| D21255.1 | 14 | 2986 | BAA04799.1 | |
| L34056.1 | 11 | 2486 | AAA35622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv572901 | Chr.16:64930330 - 64956284 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map