Assay Details
Target Gene Details
Entrez Gene ID: | 10693 |
Gene Name: | chaperonin containing TCP1 subunit 6B |
Gene Aliases: |
CCT-zeta-2, CCTZ-2, Cctz2, TCP-1-zeta-2, TSA303 |
Location: |
Chr.17:34927859-34981221 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT6B | NM_001193529.2 | NP_001180458.1 | ||
| NM_001193530.1 | NP_001180459.1 | |||
| NM_006584.3 | NP_006575.2 | |||
| XM_011524203.2 | XP_011522505.1 | |||
| XM_017024023.1 | XP_016879512.1 | |||
| XM_017024024.1 | XP_016879513.1 | |||
| AK225728.1 | ||||
| AK225729.1 | ||||
| AK301773.1 | ||||
| AK302344.1 | ||||
| BC026125.2 | AAH26125.1 | |||
| BC027591.1 | AAH27591.1 | |||
| BI825786.1 | ||||
| CR933688.1 | CAI45976.1 | |||
| D78333.1 | BAA11347.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526685 | Chr.17:34928743 - 34959645 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map