Assay Details
Target Gene Details
Entrez Gene ID: | 26121 |
Gene Name: | pre-mRNA processing factor 31 |
Gene Aliases: |
NY-BR-99, PRP31, RP11, SNRNP61 |
Location: |
Chr.19:54115410-54131719 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF31 | NM_015629.3 | NP_056444.3 | ||
| XM_006723137.3 | XP_006723200.1 | |||
| AB593024.1 | ||||
| AB593025.1 | ||||
| AF308303.1 | AAG48270.1 | |||
| AK098547.1 | BAC05329.1 | |||
| AK301709.1 | ||||
| AL050369.1 | CAB43677.1 | |||
| AY040822.1 | AAK77986.1 | |||
| BC117389.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510773 | Chr.19:54017180 - 54132236 on Build GRCh38 | Deletion |
|
| nsv515623 | Chr.19:54102901 - 54179150 on Build GRCh38 | Loss |
|
| nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion |
|
| esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
|
| nsv2545 | Chr.19:54122949 - 54142607 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map