Assay Details
Target Gene Details
Entrez Gene ID: | 7852 |
Gene Name: | C-X-C motif chemokine receptor 4 |
Gene Aliases: |
CD184, D2S201E, FB22, HM89, HSY3RR, LAP-3, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, WHIMS |
Location: |
Chr.2:136114349-136118155 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCR4 | NM_001008540.1 | 1 | 1213 | NP_001008540.1 |
| NM_003467.2 | 2 | 992 | NP_003458.1 | |
| AF025375.1 | 1 | 897 | AAB81970.1 | |
| AF147204.1 | 1 | 1251 | AAF00130.1 | |
| AF348491.1 | 2 | 940 | AAK29630.1 | |
| AK129916.1 | 2 | 986 | ||
| AK296674.1 | 3 | 885 | ||
| AK309641.1 | 1 | 1141 | ||
| AK309885.1 | 2 | 1091 | ||
| AK312244.1 | 2 | 986 | ||
| AY242129.1 | 1 | 897 | AAO92296.1 | |
| BC020968.2 | 2 | 950 | AAH20968.1 | |
| BT006660.1 | 1 | 897 | AAP35306.1 | |
| D10924.1 | 2 | 973 | BAA01722.1 | |
| KU245646.1 | 2 | 1110 | ||
| KU245647.1 | 2 | 996 | ||
| L01639.1 | 2 | 922 | AAA16594.1 | |
| L06797.1 | 2 | 969 | AAA03209.1 | |
| M99293.1 | 2 | 981 | AAA16617.1 | |
| X71635.1 | 2 | 964 | CAA50641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523997 | Chr.2:136110306 - 136463173 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map