Assay Details
Target Gene Details
Entrez Gene ID: | 9129 |
Gene Name: | pre-mRNA processing factor 3 |
Gene Aliases: |
HPRP3, HPRP3P, PRP3, Prp3p, RP18, SNRNP90 |
Location: |
Chr.1:150321479-150353233 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF3 | NM_004698.2 | NP_004689.1 | ||
| XM_011510129.1 | XP_011508431.1 | |||
| XM_011510130.2 | XP_011508432.1 | |||
| XM_011510131.2 | XP_011508433.1 | |||
| XM_011510132.2 | XP_011508434.1 | |||
| XM_017002790.1 | XP_016858279.1 | |||
| XM_017002791.1 | XP_016858280.1 | |||
| AF001947.1 | AAC09069.1 | |||
| AF016370.1 | AAC51926.1 | |||
| AK222825.1 | BAD96545.1 | |||
| AK297139.1 | ||||
| AK297265.1 | ||||
| AK299980.1 | ||||
| AK310241.1 | ||||
| AK312891.1 | ||||
| BC000184.2 | AAH00184.1 | |||
| BC001954.1 | AAH01954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831548 | Chr.1:150296285 - 150463240 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map