Assay Details
Target Gene Details
Entrez Gene ID: | 27173 |
Gene Name: | solute carrier family 39 member 1 |
Gene Aliases: |
ZIP1, ZIRTL |
Location: |
Chr.1:153959099-153968184 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A1 | NM_001271957.1 | 4 | 2162 | NP_001258886.1 |
| NM_001271958.1 | 4 | 1999 | NP_001258887.1 | |
| NM_001271959.1 | 4 | 2168 | NP_001258888.1 | |
| NM_001271960.1 | 5 | 2060 | NP_001258889.1 | |
| NM_001271961.1 | 3 | 1868 | NP_001258890.1 | |
| NM_014437.4 | 5 | 2306 | NP_055252.2 | |
| AF132942.1 | 3 | 1856 | AAD27717.1 | |
| AF151829.1 | 4 | 1877 | AAD34066.1 | |
| AJ243649.1 | 4 | 1893 | CAB59979.1 | |
| AJ271671.1 | 4 | 1907 | CAB82784.1 | |
| AK074943.1 | 4 | 2116 | ||
| AK075257.1 | 4 | 1930 | BAC11502.1 | |
| AW571711.1 | 1 | 157 | ||
| BC002563.2 | 4 | 1881 | AAH02563.1 | |
| BC003152.2 | 3 | 1853 | AAH03152.1 | |
| BC007886.2 | 4 | 1878 | AAH07886.1 | |
| BC014303.2 | 5 | 2286 | AAH14303.1 | |
| BM807264.1 | 1 | 626 |
More Information
Set Membership: |
|

Genomic Map