Assay Details
Target Gene Details
Entrez Gene ID: | 9340 |
Gene Name: | glucagon like peptide 2 receptor |
Gene Aliases: |
- |
Location: |
Chr.17:9825508-9894163 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLP2R | NM_004246.2 | 1 | 467 | NP_004237.1 |
| XM_011524077.2 | 1 | 506 | XP_011522379.1 | |
| XM_017025339.1 | 1 | 506 | XP_016880828.1 | |
| XM_017025340.1 | 1 | 510 | XP_016880829.1 | |
| XM_017025341.1 | 1 | 508 | XP_016880830.1 | |
| AF105367.1 | AAD16895.1 | |||
| AK295765.1 | ||||
| AK307283.1 | 1 | 114 | ||
| AK314269.1 | 1 | 467 | ||
| BC096261.1 | AAH96261.1 | |||
| BC096262.1 | AAH96262.1 | |||
| BC096263.1 | AAH96263.1 | |||
| BC096264.1 | AAH96264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1976 | Chr.17:9785619 - 9830655 on Build GRCh38 | Deletion |
|
| nsv833358 | Chr.17:9643743 - 9828246 on Build GRCh38 | Gain |
|
| nsv833360 | Chr.17:9734634 - 9900986 on Build GRCh38 | Loss |
|
| nsv1065401 | Chr.17:9799828 - 9986825 on Build GRCh38 | Loss |
|
| nsv833361 | Chr.17:9785863 - 9979090 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map