Assay Details
Target Gene Details
Entrez Gene ID: | 5651 |
Gene Name: | transmembrane protease, serine 15 |
Gene Aliases: |
ENTK, PRSS7 |
Location: |
Chr.21:18269107-18403806 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS15 | NM_002772.2 | 25 | 3077 | NP_002763.2 |
| XM_011529654.1 | 27 | 3351 | XP_011527956.1 | |
| XM_011529655.1 | 28 | 3269 | XP_011527957.1 | |
| XM_011529656.2 | 28 | 3276 | XP_011527958.1 | |
| XM_011529657.1 | 26 | 3306 | XP_011527959.1 | |
| XM_011529658.1 | 25 | 3270 | XP_011527960.1 | |
| XM_011529659.1 | 26 | 3241 | XP_011527961.1 | |
| BC111749.1 | 25 | 3066 | AAI11750.1 | |
| BM015908.1 | ||||
| KF688184.1 | ||||
| U09860.1 | 25 | 3086 | AAC50138.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751925 | Chr.21:18035494 - 21000411 on Build GRCh38 | Loss |
|
| nsv459105 | Chr.21:18237846 - 18282481 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map